Assay Details
Target Gene Details
Entrez Gene ID: | 53972 |
Gene Name: | neuronal guanine nucleotide exchange factor |
Gene Aliases: |
Tims2, ephexin |
Location: |
Chr.1:87476829-87573870 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ngef | NM_001111314.1 | 6 | 1171 | NP_001104784.1 |
| NM_019867.2 | 4 | 808 | NP_063920.2 | |
| AJ238898.1 | 4 | 789 | CAC00698.1 | |
| AK138433.1 | 4 | 777 | ||
| AY038025.1 | 4 | 783 | AAK71494.1 | |
| BC039279.1 | 6 | 1171 | AAH39279.1 |
More Information
Set Membership: |
|

Genomic Map