Assay Details
Target Gene Details
Entrez Gene ID: | 107895 |
Gene Name: | mannoside acetylglucosaminyltransferase 5 |
Gene Aliases: |
4930471A21Rik, 5330407H02Rik, AI480971, GlcNAc-TV |
Location: |
Chr.1:127204977-127488334 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mgat5 | NM_145128.3 | 17 | 2616 | NP_660110.2 |
| AF474154.1 | 15 | 1996 | AAL83249.1 | |
| AF474155.1 | 15 | 2059 | AAL83250.1 | |
| AK030402.1 | 17 | 2584 | BAC26946.1 | |
| BC125533.1 | 15 | 2024 | ||
| BC125535.1 | 15 | 2024 |
More Information
Set Membership: |
|

Genomic Map