Assay Details
Target Gene Details
Entrez Gene ID: | 66112 |
Gene Name: | mitochondrial amidoxime reducing component 1 |
Gene Aliases: |
1300013F15Rik, Mosc1 |
Location: |
Chr.1:184786767-184811308 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Marc1 | NM_001290273.1 | NP_001277202.1 | ||
| AK004989.1 | BAB23724.1 | |||
| AK011722.1 | ||||
| BC028441.1 | AAH28441.1 |

Genomic Map