Assay Details
Target Gene Details
Entrez Gene ID: | 67247 |
Gene Name: | mitochondrial amidoxime reducing component 2 |
Gene Aliases: |
2810484M10Rik, Mosc2, mARC1 |
Location: |
Chr.1:184813068-184846446 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Marc2 | NM_133684.3 | NP_598445.1 | ||
| AK002874.1 | ||||
| AK013440.1 | ||||
| AK017150.1 | ||||
| AK051230.1 | BAC34565.1 | |||
| BC006888.1 | AAH06888.1 |

Genomic Map