Assay Details
Target Gene Details
Entrez Gene ID: | 227157 |
Gene Name: | membrane protein, palmitoylated 4 (MAGUK p55 subfamily member 4) |
Gene Aliases: |
Dlg6 |
Location: |
Chr.1:59120935-59163389 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mpp4 | NM_001164682.1 | NP_001158154.1 | ||
| NM_145143.3 | 1 | 54 | NP_660125.2 | |
| AK044600.1 | ||||
| AK044682.1 | BAC32031.1 | |||
| BC061694.1 | ||||
| BI737565.1 | 1 | 54 |
More Information
Set Membership: |
|

Genomic Map