Assay Details
Target Gene Details
Entrez Gene ID: | 53311 |
Gene Name: | myosin binding protein H |
Gene Aliases: |
AI385643, MyBP-H |
Location: |
Chr.1:134193417-134201234 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mybph | NM_016749.2 | 2 | 349 | NP_058029.2 |
| AA673795.1 | 2 | 361 | ||
| BC116684.1 | 2 | 316 | ||
| BC118050.1 | 2 | 316 | ||
| U68267.1 | 2 | 343 | AAB08426.1 |

Genomic Map