Assay Details
Target Gene Details
Entrez Gene ID: | 59125 |
Gene Name: | NIMA (never in mitosis gene a)-related expressed kinase 7 |
Gene Aliases: |
2810460C19Rik, AU020186 |
Location: |
Chr.1:138484106-138619779 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nek7 | NM_021605.3 | NP_067618.1 | ||
| AF217650.1 | AAG16652.1 | |||
| AK035502.1 | BAC29080.1 | |||
| AK078639.1 | BAC37346.1 | |||
| AK088173.1 | BAC40190.1 | |||
| AK149308.1 | BAE28804.1 | |||
| AK165583.1 | BAE38274.1 | |||
| BC037697.1 | AAH37697.1 |
More Information
Set Membership: |
|

Genomic Map