Assay Details
Target Gene Details
Entrez Gene ID: | 11539 |
Gene Name: | adenosine A1 receptor |
Gene Aliases: |
A1-AR, A1AR, A1R, AA1R, AI848715, BB176431, Ri |
Location: |
Chr.1:134199215-134235457 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adora1 | NM_001008533.3 | 2 | 5218 | NP_001008533.1 |
| NM_001039510.2 | 3 | 4403 | NP_001034599.1 | |
| NM_001282945.1 | 3 | 4437 | NP_001269874.1 | |
| NM_001291928.1 | 2 | 3969 | NP_001278857.1 | |
| NM_001291930.1 | 2 | 4005 | NP_001278859.1 | |
| AK141531.1 | 1 | 3268 | BAE24720.1 | |
| BC079624.1 | 5 | 3988 | AAH79624.1 | |
| CF579393.1 | 1 | 620 |
Target Gene Details
Entrez Gene ID: | 53311 |
Gene Name: | myosin binding protein H |
Gene Aliases: |
AI385643, MyBP-H |
Location: |
Chr.1:134193417-134201234 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mybph | NM_016749.2 | NP_058029.2 | ||
| AK029449.1 | ||||
| BC116684.1 | ||||
| BC118050.1 | ||||
| CB587371.1 | ||||
| U68267.1 | AAB08426.1 |
More Information
Set Membership: |
|

Genomic Map