Assay Details
Target Gene Details
Entrez Gene ID: | 215690 |
Gene Name: | neuron navigator 1 |
Gene Aliases: |
9530089B19, 9930003A20Rik, C230080M11Rik, Pomfil3, Unc53h1, mKIAA1151, steerin-1 |
Location: |
Chr.1:135434580-135688116 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nav1 | NM_173437.2 | 15 | 3491 | NP_775613.2 |
| AF307453.1 | 15 | 3851 | AAO13290.1 | |
| AK053255.1 | 13 | 2470 | BAC35323.1 | |
| AK082667.1 | 13 | 2404 | BAC38568.1 | |
| AK122458.1 | 16 | 3558 | BAC65740.1 | |
| AK140435.1 | ||||
| BC059840.1 | 15 | 3634 | AAH59840.1 | |
| CN455316.1 | 6 | 476 |

Genomic Map