Assay Details
Target Gene Details
Entrez Gene ID: | 269198 |
Gene Name: | neurobeachin like 1 |
Gene Aliases: |
2310076G13Rik, A530050O19Rik, A530083I02Rik, AI662228, ALS2CR17 |
Location: |
Chr.1:60178629-60334705 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 14 - Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nbeal1 | NM_173444.2 | NP_775620.2 | ||
| AK040951.1 | BAC30758.1 | |||
| AK143706.1 |

Genomic Map