Assay Details
Target Gene Details
Entrez Gene ID: | 11545 |
Gene Name: | poly (ADP-ribose) polymerase family, member 1 |
Gene Aliases: |
5830444G22Rik, AI893648, ARTD1, Adprp, Adprt1, C80510, PARP, PPOL, parp-1, sPARP-1 |
Location: |
Chr.1:180568975-180601254 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Parp1 | NM_007415.2 | 7 | 1026 | NP_031441.2 |
| AK028042.1 | 7 | 1045 | ||
| AK030889.1 | 7 | 1029 | BAC27173.1 | |
| AK088679.1 | 7 | 1030 | BAC40500.1 | |
| AK146677.1 | 7 | 1047 | BAE27352.1 | |
| AK152360.1 | 7 | 1047 | BAE31150.1 | |
| AK159433.1 | 7 | 1078 | BAE35080.1 | |
| AK165158.1 | 7 | 1046 | ||
| AK166104.1 | 7 | 1078 | BAE38574.1 | |
| AK167699.1 | 7 | 1018 | BAE39743.1 | |
| AK169326.1 | 7 | 1047 | BAE41080.1 | |
| AK210311.1 | ||||
| BC012041.1 | 7 | 1026 | AAH12041.1 | |
| X14206.1 | 7 | 1048 | CAA32421.1 |
More Information
Set Membership: |
|

Genomic Map