Assay Details
Target Gene Details
Entrez Gene ID: | 26424 |
Gene Name: | nuclear receptor subfamily 5, group A, member 2 |
Gene Aliases: |
AU020803, D1Ertd308e, Ftf, LRH-1, UF2-H3B |
Location: |
Chr.1:136842562-136960448 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nr5a2 | NM_001159769.2 | 1 | 76 | NP_001153241.1 |
| NM_030676.3 | NP_109601.1 | |||
| AB221629.1 | ||||
| BB652042.1 | ||||
| BC137845.1 | ||||
| DQ133942.1 | 1 | 76 | AAZ99450.1 | |
| M81385.1 | AAA39446.1 |
More Information
Set Membership: |
|

Genomic Map