Assay Details
Target Gene Details
Entrez Gene ID: | 75584 |
Gene Name: | RIKEN cDNA 2310030A07 gene |
Gene Aliases: |
- |
Location: |
Chr.1:150320681-150346771 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 2310030A07Rik | NR_040603.1 | |||
| AK009534.1 |
Target Gene Details
Entrez Gene ID: | 20028 |
Gene Name: | phosducin |
Gene Aliases: |
Rpr-1, Rpr1 |
Location: |
Chr.1:150319417-150333906 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pdc | NM_001159730.1 | NP_001153202.1 | ||
| NM_024458.2 | NP_077778.1 | |||
| AK020798.1 | ||||
| BC012507.1 | AAH12507.1 | |||
| BC021920.1 | AAH21920.1 | |||
| BI738229.1 | ||||
| L08075.1 | AAK13559.1 |
More Information
Set Membership: |
|

Genomic Map