Assay Details
Target Gene Details
Entrez Gene ID: | 381318 |
Gene Name: | NSL1, MIND kinetochore complex component, homolog (S. cerevisiae) |
Gene Aliases: |
4833432M17Rik, AI451597, Gm105, Gm984 |
Location: |
Chr.1:191063012-191084558 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nsl1 | NM_198654.3 | 3 | 428 | NP_941056.3 |
| AK029421.1 | 3 | 426 | BAC26444.1 | |
| BC029086.1 | 3 | 418 | AAH29086.1 |

Genomic Map