Assay Details
Target Gene Details
Entrez Gene ID: | 241113 |
Gene Name: | protein kinase, AMP-activated, gamma 3 non-catatlytic subunit |
Gene Aliases: |
AMPKg3L, AMPKg3S, Amkg3, Ampkg3 |
Location: |
Chr.1:74738922-74749590 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prkag3 | NM_153744.3 | 9 | 1048 | NP_714966.1 |
| AF525500.1 | 9 | 955 | AAN47137.1 | |
| AF525501.1 | 8 | 976 | AAN47138.1 | |
| AK036585.1 | 9 | 1049 | BAC29492.1 | |
| BC116749.1 | 8 | 912 | ||
| BC116777.1 | 8 | 912 |
More Information
Set Membership: |
|

Genomic Map