Assay Details
Target Gene Details
Entrez Gene ID: | 77300 |
Gene Name: | Ras association (RalGDS/AF-6) and pleckstrin homology domains 1 |
Gene Aliases: |
9430025M21Rik, C730009O10Rik, Lpd |
Location: |
Chr.1:60483185-60567074 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Raph1 | NM_001045513.3 | 6 | 1098 | NP_001038978.3 |
| AA199065.1 | 5 | 496 | ||
| CB246004.1 | 3 | 629 |
More Information
Set Membership: |
|

Genomic Map