Assay Details
Target Gene Details
Entrez Gene ID: | 20215 |
Gene Name: | S-antigen, retina and pineal gland (arrestin) |
Gene Aliases: |
A930001K18Rik, Arr1, Irbp, arrestin |
Location: |
Chr.1:87803680-87845158 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sag | NM_009118.2 | NP_033144.1 | ||
| AK044214.1 | BAC31820.1 | |||
| AK053278.1 | BAC35331.1 | |||
| AK143098.1 | BAE25268.1 | |||
| AK149299.1 | BAE28798.1 | |||
| AY651760.1 | AAV54097.1 | |||
| BC016498.1 | AAH16498.1 | |||
| M24086.1 | AAA40090.1 |
More Information
Set Membership: |
|

Genomic Map