Assay Details
Target Gene Details
Entrez Gene ID: | 212712 |
Gene Name: | special AT-rich sequence binding protein 2 |
Gene Aliases: |
mKIAA1034 |
Location: |
Chr.1:56793981-56978681 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Satb2 | NM_139146.2 | 10 | 1877 | NP_631885.1 |
| AF319623.1 | 10 | 1877 | AAL37172.1 | |
| AJ606045.1 | 9 | 1595 | CAE54289.1 | |
| AK129270.1 | 9 | 2868 | BAC98080.1 | |
| BC138625.1 | 9 | 1628 | ||
| BC138626.1 | 9 | 1628 |
More Information
Set Membership: |
|

Genomic Map