Assay Details
Target Gene Details
Entrez Gene ID: | 320139 |
Gene Name: | protein tyrosine phosphatase, non-receptor type 7 |
Gene Aliases: |
BPTP-4, C920001D21Rik, HEPTP, LC-PTP, LPTP |
Location: |
Chr.1:135131840-135145328 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ptpn7 | NM_177081.3 | 3 | 369 | NP_796055.1 |
| AK041415.1 | 1 | 1262 | ||
| AK049175.1 | 2 | 220 | BAC33587.1 | |
| AK083305.1 | 3 | 369 | BAC38856.1 | |
| BC098475.1 | 3 | 423 | AAH98475.1 | |
| BC138194.1 | 2 | 305 | ||
| BC145190.1 | 2 | 305 |

Genomic Map