Assay Details
Target Gene Details
Entrez Gene ID: | 12421 |
Gene Name: | RB1-inducible coiled-coil 1 |
Gene Aliases: |
2900055E04Rik, 5930404L04Rik, Cc1, FIP200, LaXp180 |
Location: |
Chr.1:6214642-6276372 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 16 - Intron 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rb1cc1 | NM_009826.4 | NP_033956.2 | ||
| AB050017.2 | BAB16846.2 | |||
| AB070619.1 | BAB85610.1 | |||
| AB093216.1 | BAC41400.1 | |||
| AK135130.1 | ||||
| BC150774.1 |
More Information
Set Membership: |
|

Genomic Map