Assay Details
Target Gene Details
Entrez Gene ID: | 214742 |
Gene Name: | REST corepressor 3 |
Gene Aliases: |
4921514E24Rik, C730034D20Rik, E130101E15Rik |
Location: |
Chr.1:192098546-192138067 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rcor3 | NM_001290278.1 | 5 | 536 | NP_001277207.1 |
| NM_001290279.1 | 5 | 536 | NP_001277208.1 | |
| NM_001290280.1 | 5 | 536 | NP_001277209.1 | |
| NM_001290282.1 | 6 | 650 | NP_001277211.1 | |
| NM_144814.3 | 5 | 536 | NP_659063.3 | |
| AK040839.1 | 5 | 523 | ||
| AK050287.1 | 6 | 641 | ||
| AK135612.1 | 5 | 509 | ||
| AK155440.1 | 5 | 508 | ||
| AK193623.1 | ||||
| AK203210.1 | ||||
| BC057141.1 | 5 | 283 | ||
| BC138938.1 | 5 | 256 | ||
| BC138939.1 | 5 | 256 |

Genomic Map