Assay Details
Target Gene Details
Entrez Gene ID: | 226594 |
Gene Name: | RCSD domain containing 1 |
Gene Aliases: |
A430105K13Rik, BC025872 |
Location: |
Chr.1:165648945-165710063 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rcsd1 | NM_001038846.1 | NP_001033935.1 | ||
| NM_178593.3 | NP_848708.2 | |||
| AK133961.1 | BAE21956.1 | |||
| AK158429.1 | BAE34505.1 | |||
| AK168816.1 | BAE40643.1 | |||
| AK171914.1 | BAE42730.1 | |||
| BC025872.1 | AAH25872.1 | |||
| BY230131.1 |
More Information
Set Membership: |
|

Genomic Map