Assay Details
Target Gene Details
Entrez Gene ID: | 19888 |
Gene Name: | retinitis pigmentosa 1 (human) |
Gene Aliases: |
Dcdc3, Orp1, Rp1h, mG145 |
Location: |
Chr.1:4290846-4409266 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rp1 | NM_001195662.1 | 3 | 749 | NP_001182591.1 |
| NM_011283.2 | 3 | 792 | NP_035413.1 | |
| AF155141.2 | 3 | 783 | AAD42089.2 | |
| AK044413.1 | 3 | 785 | BAC31909.1 | |
| AK141793.1 | 3 | 751 | ||
| BC096607.1 | 3 | 792 | AAH96607.1 | |
| BC120927.1 | 3 | 746 | ||
| BC120928.1 | 3 | 746 | ||
| BM937589.1 | 1 | 121 |

Genomic Map