Assay Details
Target Gene Details
Entrez Gene ID: | 81898 |
Gene Name: | splicing factor 3b, subunit 1 |
Gene Aliases: |
155kDa, 2810001M05Rik, AA409119, Prp10, SAP155, SF3b155, TA-8, Targ4 |
Location: |
Chr.1:54985169-55027500 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sf3b1 | NM_031179.2 | NP_112456.2 | ||
| AB037890.1 | BAB40140.1 | |||
| AK012632.1 | BAB28369.1 | |||
| AK140982.1 | BAE24537.1 | |||
| AK146989.1 | BAE27592.1 | |||
| AK165992.1 | ||||
| AK171454.1 | BAE42463.1 | |||
| BC003419.1 | ||||
| BC028959.1 | ||||
| CK331410.1 |
More Information
Set Membership: |
|

Genomic Map