Assay Details
Target Gene Details
Entrez Gene ID: | 240880 |
Gene Name: | SCY1-like 3 (S. cerevisiae) |
Gene Aliases: |
1200016D23Rik, 6030457O16, AW214499, Pace1 |
Location: |
Chr.1:163927958-163955126 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Scyl3 | NM_001286002.1 | NP_001272931.1 | ||
| NM_001286003.1 | NP_001272932.1 | |||
| NM_028776.5 | NP_083052.1 | |||
| AK004809.2 | BAB23580.1 | |||
| AK030824.1 | ||||
| AK044100.1 | BAC31777.1 | |||
| AK162937.1 | BAE37123.1 | |||
| BC043085.1 | AAH43085.1 |
More Information
Set Membership: |
|

Genomic Map