Assay Details
Target Gene Details
Entrez Gene ID: | 545378 |
Gene Name: | SH2 domain containing 1B2 |
Gene Aliases: |
EAT-2B, Eat2b, Ert, Sh2d1c |
Location: |
Chr.1:170232871-170251587 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sh2d1b2 | NM_001033499.1 | NP_001028671.1 | ||
| DQ131181.1 | ||||
| DQ140178.1 | AAZ94715.1 |

Genomic Map