Assay Details
Target Gene Details
Entrez Gene ID: | 89867 |
Gene Name: | SEC16 homolog B (S. cerevisiae) |
Gene Aliases: |
Lztr2, Rgpr, Rgpr-p117 |
Location: |
Chr.1:157506733-157568425 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 18 - Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sec16b | NM_001159986.1 | NP_001153458.1 | ||
| NM_033354.3 | NP_203505.3 | |||
| NR_027641.1 | ||||
| AB063356.1 | BAB61034.1 | |||
| AK132144.1 | ||||
| AK173302.1 | BAD32580.1 | |||
| BC059194.1 | AAH59194.1 |

Genomic Map