Assay Details
Target Gene Details
Entrez Gene ID: | 80721 |
Gene Name: | solute carrier family 19, member 3 |
Gene Aliases: |
A230084E24Rik, AI788884, ThTr2 |
Location: |
Chr.1:83012523-83038859 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc19a3 | NM_030556.2 | 5 | 1237 | NP_085033.2 |
| AF271634.1 | 5 | 1237 | AAG53880.1 | |
| AK079566.1 | 2 | 156 | ||
| AK098112.1 | 5 | 1228 | ||
| BC109154.1 | 5 | 1220 | AAI09155.1 | |
| BC109155.1 | 4 | 1090 | AAI09156.1 |
More Information
Set Membership: |
|

Genomic Map