Assay Details
Target Gene Details
Entrez Gene ID: | 18000 |
Gene Name: | septin 2 |
Gene Aliases: |
AW208991, Nedd-5, Nedd5, mKIAA0158 |
Location: |
Chr.1:93478967-93509733 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sept2 | NM_001159717.1 | NP_001153189.1 | ||
| NM_001159718.1 | NP_001153190.1 | |||
| NM_001159719.1 | NP_001153191.1 | |||
| NM_010891.2 | NP_035021.1 | |||
| AK028072.1 | BAC25737.1 | |||
| AK146616.1 | BAE27305.1 | |||
| AK151591.1 | BAE30531.1 | |||
| AK169852.1 | ||||
| AK171331.1 | BAE42396.1 | |||
| BC138636.1 | ||||
| BC138637.1 | ||||
| D49382.2 | BAA08380.1 |
More Information
Set Membership: |
|

Genomic Map