Assay Details
Target Gene Details
Entrez Gene ID: | 20846 |
Gene Name: | signal transducer and activator of transcription 1 |
Gene Aliases: |
2010005J02Rik, AA408197 |
Location: |
Chr.1:52119438-52161865 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Stat1 | NM_001205314.1 | 1 | 95 | NP_001192243.1 |
| NM_009283.4 | 1 | 95 | NP_033309.3 | |
| AK018544.1 | BAB31265.1 | |||
| AK039458.1 | 1 | 96 | BAC30355.1 | |
| AK041599.1 | BAE43313.1 | |||
| AK041814.1 | BAE43314.1 | |||
| AK082706.1 | 1 | 77 | BAC38579.1 | |
| AK084855.1 | BAC39293.1 | |||
| AK159887.1 | 1 | 79 | BAE35456.1 | |
| BC004808.1 | AAH04808.1 |
More Information
Set Membership: |
|

Genomic Map