Assay Details
Target Gene Details
Entrez Gene ID: | 68428 |
Gene Name: | STEAP family member 3 |
Gene Aliases: |
1010001D01Rik, Tsap6, pHyde |
Location: |
Chr.1:120226414-120271082 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Steap3 | NM_001085409.1 | NP_001078878.1 | ||
| AK167028.1 | BAE39201.1 |

Genomic Map