Assay Details
Target Gene Details
Entrez Gene ID: | 68428 |
Gene Name: | STEAP family member 3 |
Gene Aliases: |
1010001D01Rik, Tsap6, pHyde |
Location: |
Chr.1:120226414-120271082 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Steap3 | NM_001085409.1 | 5 | 1542 | NP_001078878.1 |
| NM_133186.3 | 4 | 1169 | NP_573449.2 | |
| AK028961.1 | 4 | 1146 | ||
| AK078769.1 | 4 | 1563 | BAC37383.1 | |
| AK167028.1 | 5 | 1542 | BAE39201.1 | |
| AK171237.1 | 4 | 1174 | BAE42333.1 | |
| AK217925.1 | 2 | 248 | ||
| AY029586.1 | 4 | 1157 | AAK50539.1 | |
| BC037435.1 | 4 | 1170 | AAH37435.1 |

Genomic Map