Assay Details
Target Gene Details
Entrez Gene ID: | 71728 |
Gene Name: | serine/threonine kinase 11 interacting protein |
Gene Aliases: |
1200014D22Rik, BB131189, LIP1, LKB1IP |
Location: |
Chr.1:75521529-75537335 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Stk11ip | NM_027886.3 | 21 | 2717 | NP_082162.3 |
| AK004757.2 | 21 | 2670 | BAB23538.1 | |
| AK156122.1 | 20 | 2805 | BAE33593.1 | |
| AK165911.1 | 21 | 2710 | ||
| AK171989.1 | 20 | 2579 | BAE42762.1 | |
| BC061635.1 | 21 | 2700 | AAH61635.1 |
More Information
Set Membership: |
|

Genomic Map