Assay Details
Target Gene Details
Entrez Gene ID: | 20668 |
Gene Name: | SRY (sex determining region Y)-box 13 |
Gene Aliases: |
AA407916, AW259412, AW540933, Sox-13 |
Location: |
Chr.1:133382300-133424212 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sox13 | NM_011439.2 | 5 | 824 | NP_035569.2 |
| AB006329.1 | 4 | 480 | BAA25786.1 | |
| AJ000740.1 | 4 | 434 | CAA04278.1 | |
| BC007130.1 | 5 | 646 | AAH07130.1 |

Genomic Map