Assay Details
Target Gene Details
Entrez Gene ID: | 59041 |
Gene Name: | serine/threonine kinase 25 (yeast) |
Gene Aliases: |
1500019J11Rik, AU018434, C86992, Ste20-like, Ysk1 |
Location: |
Chr.1:93620751-93654216 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Stk25 | NM_021537.3 | 9 | 1211 | NP_067512.3 |
| AF004934.1 | 8 | 1049 | AAD01208.1 | |
| AK005289.1 | 1 | 69 | ||
| AK148041.1 | 9 | 1185 | BAE28307.1 | |
| AK152917.1 | 9 | 1154 | BAE31594.1 | |
| BC052913.1 | 9 | 1146 | AAH52913.1 | |
| BC071218.1 | 9 | 1206 | AAH71218.1 |

Genomic Map