Assay Details
Target Gene Details
Entrez Gene ID: | 21346 |
Gene Name: | transgelin 2 |
Gene Aliases: |
2700094C18Rik, SM22beta, Sm22B, Sm22a |
Location: |
Chr.1:172500246-172507375 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tagln2 | NM_178598.2 | NP_848713.1 | ||
| AF149291.1 | AAD37787.1 | |||
| AF465519.1 | AAM54133.1 | |||
| AK151576.1 | BAE30516.1 | |||
| AK167395.1 | BAE39484.1 | |||
| AK167922.1 | BAE39928.1 | |||
| AK171088.1 | BAE42241.1 | |||
| AK172892.1 | BAD32170.1 | |||
| AK195747.1 | ||||
| BC049861.1 |
More Information
Set Membership: |
|

Genomic Map