Assay Details
Target Gene Details
Entrez Gene ID: | 68972 |
Gene Name: | TatD DNase domain containing 3 |
Gene Aliases: |
1500010M24Rik |
Location: |
Chr.1:191045826-191063010 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tatdn3 | NM_001163421.1 | NP_001156893.1 | ||
| NM_001163422.1 | NP_001156894.1 | |||
| NM_026895.1 | NP_081171.1 | |||
| AK005193.1 | BAB23875.1 | |||
| AK156071.1 | BAE33572.1 | |||
| BC061248.1 | AAH61248.1 |
More Information
Set Membership: |
|

Genomic Map