Assay Details
Target Gene Details
Entrez Gene ID: | 30935 |
Gene Name: | torsin family 3, member A |
Gene Aliases: |
Adir |
Location: |
Chr.1:156653617-156674339 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tor3a | NM_023141.2 | NP_075630.2 | ||
| AJ299404.1 | CAC13978.1 | |||
| AK007814.1 | BAB25279.1 | |||
| AK009693.1 | BAB26443.1 | |||
| AK017908.1 | BAB30998.1 | |||
| AK151048.1 | BAE30065.1 | |||
| AK151390.1 | ||||
| AK152243.1 | BAE31066.1 | |||
| AK152358.1 | BAE31149.1 | |||
| AK152484.1 | BAE31256.1 | |||
| AK153391.1 | BAE31955.1 | |||
| AK170296.1 | BAE41695.1 | |||
| AK172355.1 | BAE42964.1 | |||
| BC071216.1 | AAH71216.1 |
More Information
Set Membership: |
|

Genomic Map