Assay Details
Target Gene Details
Entrez Gene ID: | 30935 |
Gene Name: | torsin family 3, member A |
Gene Aliases: |
Adir |
Location: |
Chr.1:156653617-156674339 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tor3a | NM_023141.2 | 5 | 880 | NP_075630.2 |
| AJ299404.1 | 5 | 848 | CAC13978.1 | |
| AK007814.1 | 5 | 869 | BAB25279.1 | |
| AK009693.1 | 5 | 898 | BAB26443.1 | |
| AK017908.1 | 5 | 825 | BAB30998.1 | |
| AK151048.1 | 5 | 866 | BAE30065.1 | |
| AK151390.1 | 6 | 917 | ||
| AK152243.1 | 5 | 845 | BAE31066.1 | |
| AK152358.1 | 5 | 881 | BAE31149.1 | |
| AK152484.1 | 5 | 881 | BAE31256.1 | |
| AK153391.1 | 5 | 866 | BAE31955.1 | |
| AK170296.1 | 5 | 875 | BAE41695.1 | |
| AK172355.1 | 5 | 855 | BAE42964.1 | |
| BC071216.1 | 5 | 861 | AAH71216.1 |
More Information
Set Membership: |
|

Genomic Map