Assay Details
Target Gene Details
Entrez Gene ID: | 226591 |
Gene Name: | TIP41, TOR signalling pathway regulator-like (S. cerevisiae) |
Gene Aliases: |
1810011K17Rik |
Location: |
Chr.1:165212286-165282478 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tiprl | NM_145513.4 | NP_663488.1 | ||
| AK049119.1 | BAC33553.1 | |||
| AK049784.1 | BAC33919.1 | |||
| AK075774.1 | BAC35948.1 | |||
| BC002098.1 | AAH02098.1 | |||
| BC058250.1 | AAH58250.1 | |||
| CF531065.1 |

Genomic Map