Assay Details
Target Gene Details
Entrez Gene ID: | 240873 |
Gene Name: | tumor necrosis factor (ligand) superfamily, member 18 |
Gene Aliases: |
Gitrl |
Location: |
Chr.1:161494657-161505290 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tnfsf18 | NM_183391.3 | 1 | 101 | NP_899247.3 |
| AJ577579.1 | 1 | 103 | CAE12166.1 | |
| AY234223.1 | 1 | 59 | AAO89011.1 | |
| AY267900.1 | AAP96745.1 | |||
| AY320040.1 | 1 | 51 | AAP70494.1 | |
| AY359852.1 | AAQ55265.1 | |||
| BC137814.1 | 1 | 101 | ||
| BC137815.1 | 1 | 101 |

Genomic Map