Assay Details
Target Gene Details
Entrez Gene ID: | 21952 |
Gene Name: | troponin I, skeletal, slow 1 |
Gene Aliases: |
2700018B22Rik, AI747285, ssTnI |
Location: |
Chr.1:135783035-135810989 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tnni1 | NM_001112702.1 | 9 | 605 | NP_001106173.1 |
| NM_021467.5 | 9 | 653 | NP_067442.1 | |
| AA230582.1 | ||||
| AA271187.1 | 9 | 604 | ||
| AJ242874.1 | 9 | 653 | CAB48403.1 | |
| AK012258.1 | 9 | 636 | BAB28123.1 | |
| BC023170.1 | 8 | 717 | AAH23170.1 |
More Information
Set Membership: |
|

Genomic Map