Assay Details
Target Gene Details
Entrez Gene ID: | 22789 |
Gene Name: | zona pellucida 3 receptor |
Gene Aliases: |
AV264399, SP56 |
Location: |
Chr.1:130544620-130629634 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zp3r | NM_009581.2 | 7 | 791 | NP_033607.2 |
| BC119121.1 | 7 | 781 | ||
| BC119123.1 | 7 | 781 | ||
| BU962248.1 | 4 | 371 | ||
| U17108.1 | 7 | 789 | AAC52208.1 |

Genomic Map