Assay Details
Target Gene Details
Entrez Gene ID: | 98496 |
Gene Name: | phosphotyrosine interaction domain containing 1 |
Gene Aliases: |
5033414K04Rik, NYGGF4 |
Location: |
Chr.1:84036293-84284675 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pid1 | NM_001003948.2 | 1 | 262 | NP_001003948.2 |
| AK035542.1 | 1 | 137 | ||
| AK083256.1 | 1 | 182 | ||
| AK150971.1 | 1 | 200 | BAE30002.1 | |
| AK151267.1 | 1 | 232 | BAE30254.1 | |
| AK163691.1 | 1 | 241 | ||
| AK164328.1 | 1 | 229 | BAE37740.1 | |
| BC080290.1 | 1 | 208 | AAH80290.1 | |
| BY004446.1 | 1 | 262 |

Genomic Map