Assay Details
Target Gene Details
Entrez Gene ID: | 226977 |
Gene Name: | ARP1 actin-related protein 1B, centractin beta |
Gene Aliases: |
2310066K23Rik, AA960180, AI851923, Arp1b |
Location: |
Chr.1:36698114-36710017 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Actr1b | NM_146107.2 | NP_666219.1 | ||
| AK078508.1 | ||||
| AK088839.1 | BAC40604.1 | |||
| AK180123.1 | ||||
| AK201389.1 | ||||
| AK205617.1 | ||||
| BC023020.1 | AAH23020.1 |
More Information
Set Membership: |
|

Genomic Map