Assay Details
Target Gene Details
Entrez Gene ID: | 319695 |
Gene Name: | ankyrin and armadillo repeat containing |
Gene Aliases: |
4932422E22Rik |
Location: |
Chr.1:72642980-72700579 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ankar | NM_176980.4 | 10 | 2248 | NP_795954.2 |
| AK030065.1 | 10 | 2249 | BAC26765.1 | |
| AK077036.1 | 9 | 1637 | ||
| BC132415.1 | 10 | 2223 | ||
| BC145253.1 | 10 | 2220 |

Genomic Map