Assay Details
Target Gene Details
Entrez Gene ID: | 12912 |
Gene Name: | cAMP responsive element binding protein 1 |
Gene Aliases: |
2310001E10Rik, 3526402H21Rik, AV083133, Creb, Creb-1 |
Location: |
Chr.1:64532804-64604548 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Creb1 | NM_001037726.1 | 1 | 140 | NP_001032815.1 |
NM_009952.2 | 1 | 140 | NP_034082.1 | |
NM_133828.2 | 1 | 140 | NP_598589.2 | |
AF448507.1 | AAL47130.1 | |||
AK042595.1 | 1 | 113 | ||
AK079223.1 | 1 | 136 | ||
BC021649.1 | 1 | 73 | AAH21649.1 | |
BY315899.1 | 1 | 150 | ||
BY782293.1 | 1 | 97 | ||
M95106.1 | 1 | 50 | AAA37456.1 | |
X92497.1 | 1 | 117 | CAA63242.1 |
