Assay Details
Target Gene Details
Entrez Gene ID: | 110611 |
Gene Name: | high density lipoprotein (HDL) binding protein |
Gene Aliases: |
1110005P14Rik, AA960365, AI118566, D1Ertd101e |
Location: |
Chr.1:93405940-93478917 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hdlbp | NM_001301364.1 | 18 | 2499 | NP_001288293.1 |
| NM_133808.5 | 18 | 2582 | NP_598569.1 | |
| AK132355.1 | 18 | 2485 | BAE21122.1 | |
| AK153965.1 | 18 | 2448 | BAE32283.1 | |
| AK161663.1 | 18 | 2451 | ||
| BC021765.1 | 17 | 2361 | AAH21765.1 | |
| BC023806.1 | 18 | 2463 | AAH23806.1 | |
| BC025648.1 | 18 | 2504 | AAH25648.1 | |
| BC027067.1 | 9 | 1000 | ||
| BC027779.1 | 18 | 2477 | AAH27779.1 | |
| BC027788.1 | 18 | 2477 | AAH27788.1 | |
| BC035301.1 | 17 | 2390 | AAH35301.1 |
More Information
Set Membership: |
|

Genomic Map