Assay Details
Target Gene Details
Entrez Gene ID: | 110611 |
Gene Name: | high density lipoprotein (HDL) binding protein |
Gene Aliases: |
1110005P14Rik, AA960365, AI118566, D1Ertd101e |
Location: |
Chr.1:93405940-93478917 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hdlbp | NM_001301364.1 | 6 | 822 | NP_001288293.1 |
| NM_133808.5 | 6 | 905 | NP_598569.1 | |
| AK048780.1 | 6 | 774 | BAC33456.1 | |
| AK132355.1 | 6 | 808 | BAE21122.1 | |
| AK153965.1 | 6 | 771 | BAE32283.1 | |
| AK161663.1 | 6 | 774 | ||
| AK186084.1 | 2 | 181 | ||
| AK187596.1 | 1 | 125 | ||
| AK188129.1 | 2 | 181 | ||
| AK201377.1 | 1 | 100 | ||
| BC021765.1 | 5 | 684 | AAH21765.1 | |
| BC023806.1 | 6 | 786 | AAH23806.1 | |
| BC025648.1 | 6 | 827 | AAH25648.1 | |
| BC027779.1 | 6 | 800 | AAH27779.1 | |
| BC027788.1 | 6 | 800 | AAH27788.1 | |
| BC035301.1 | 5 | 713 | AAH35301.1 |
More Information
Set Membership: |
|

Genomic Map