Assay Details
Target Gene Details
Entrez Gene ID: | 12290 |
Gene Name: | calcium channel, voltage-dependent, R type, alpha 1E subunit |
Gene Aliases: |
A430040I15, BII, Cach6, Cacnl1a6, Cav2.3, Cchra1, alpha1E |
Location: |
Chr.1:154390806-154884782 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 29 - Exon 30 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cacna1e | NM_009782.3 | NP_033912.2 | ||
| BC150750.1 |
More Information
Set Membership: |
|

Genomic Map